missing translation for 'onlineSavingsMsg'
Learn More
Ethylxanthic acid potassium salt, 97+%, Thermo Scientific Chemicals Product Code.: 10568702

Ethylxanthic acid potassium salt, 97+%, Thermo Scientific Chemicals

Product Code. 10568702
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10568702

Brand: Thermo Scientific Acros 410500250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-89-6
Molecular Formula C3H5KOS2
Molecular Weight (g/mol) 160.29
MDL Number MFCD00004931
InChI Key JCBJVAJGLKENNC-UHFFFAOYSA-M
Synonym potassium ethylxanthate, potassium ethyl xanthate, ethylxanthic acid potassium salt, potassium xanthogenate, potassium xanthate, potassium o-ethyl carbonodithioate, potassium ethyl xanthogenate, potassium ethylxanthogenate, potassium o-ethyl dithiocarbonate, z 3 pesticide
PubChem CID 2735045
IUPAC Name potassium;ethoxymethanedithioate
SMILES [K+].CCOC([S-])=S

Specifications

Melting Point 209°C to 214°C
Color Yellow
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula CH3CH2OC(S)SK
Quantity 25 g
Merck Index 15, 7813
Solubility Information Solubility in water: soluble
Formula Weight 160.3
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material Ethylxanthic acid potassium salt
Show More Show Less
compliance-icons
Product Identifier
  • Ethylxanthic acid potassium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylxanthic acid potassium salt, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.