Learn More
Ethynyl Estradiol, MedChemExpress
MedChemExpress Ethynyl Estradiol (17α-Ethynylestradiol;Ethynylestradiol) is an orally bio-active estrogen used in almost all modern formulations of combined oral contraceptive pills.
Brand: Medchem Express HY-B0216
This item is not returnable.
View return policy
Description
Ethynyl Estradiol (17α-EthynylestradiolSpecifications
Ethynyl Estradiol | |
White | |
C20H24O2 | |
17α-Ethynylestradiol Ethynylestradiol | |
OC1=CC=C2C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4(C#C)O)=C1 | |
296.4 | |
Research | |
Cancer-programmed cell death |
57-63-6 | |
Research | |
100 mg | |
DMSO : 250 mg/mL (843.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
296.4 | |
99.28% | |
Solid |
Research purposes only