missing translation for 'onlineSavingsMsg'
Learn More

Ethynylmagnesium chloride, 0.5M solution in THF/Toluene, AcroSeal™, Thermo Scientific Chemicals

66.50 € - 240.00 €

Chemical Identifiers

CAS 65032-27-1, 108-88-3, 109-99-9
Molecular Formula C2HClMg
Molecular Weight (g/mol) 84.79
MDL Number MFCD00075343
InChI Key YEWNLGTVYUADOR-UHFFFAOYSA-M
Synonym ethynylmagnesium chloride, ethynylmagnesiumchloride, ethynylchloromagnesium, ethynyl magnesium chloride, ethynyl-magnesium chloride, chloro ethynyl magnesium, yewnlgtvyuador-uhfffaoysa-m, ethynylmagnesium chloride solution, 0.5 m in thf
PubChem CID 2734899
IUPAC Name chloro(ethynyl)magnesium
SMILES Cl[Mg]C#C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10629204
View Documents
Thermo Scientific Acros
388951000
100 mL AcroSeal™ Glass bottle
66.50 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10085792
View Documents
Thermo Scientific Acros
388958000
800 mL AcroSeal™ Glass Bottle
240.00 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

65032-27-1, 108-88-3, 109-99-9
84.79
YEWNLGTVYUADOR-UHFFFAOYSA-M
2734899
Cl[Mg]C#C
C2HClMg
MFCD00075343
ethynylmagnesium chloride, ethynylmagnesiumchloride, ethynylchloromagnesium, ethynyl magnesium chloride, ethynyl-magnesium chloride, chloro ethynyl magnesium, yewnlgtvyuador-uhfffaoysa-m, ethynylmagnesium chloride solution, 0.5 m in thf
chloro(ethynyl)magnesium

Specifications

Brown
C2HClMg
MFCD00075343
0.9600g/mL
Solubility in water: reacts.
Cl[Mg]C#C
84.79
84.79
Liquid
65032-27-1, 108-88-3, 109-99-9
HC≡CMgCl
4 to 6%
ethynylmagnesium chloride, ethynylmagnesiumchloride, ethynylchloromagnesium, ethynyl magnesium chloride, ethynyl-magnesium chloride, chloro ethynyl magnesium, yewnlgtvyuador-uhfffaoysa-m, ethynylmagnesium chloride solution, 0.5 m in thf
YEWNLGTVYUADOR-UHFFFAOYSA-M
chloro(ethynyl)magnesium
2734899
0.96
Ethynylmagnesium chloride, 0.6M solution in THF/Toluene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of damaging the unborn child. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

Recommended Storage : Product tends to crystallize at lower temperatures. Store above 15°C. Precipitates can be redissolved by warming and appropriate homogenization.

Product Identifier
  • Ethynylmagnesium chloride, 0.6M solution in THF/toluene

Signal Word
  • Danger

Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only