missing translation for 'onlineSavingsMsg'
Learn More

Ethynyltri-n-butyltin, 96%, Thermo Scientific Chemicals

62.91 € - 203.01 €

Chemical Identifiers

CAS 994-89-8
Molecular Formula C14H28Sn
Molecular Weight (g/mol) 315.088
MDL Number MFCD00009420
InChI Key YEMJHNYABQHWHL-UHFFFAOYSA-N
Synonym tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
PubChem CID 621176
IUPAC Name tributyl(ethynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339006
View Documents
Thermo Scientific Alfa Aesar
L20178.03
1 g
62.91 €
1g
Please to purchase this item. Need a web account? Register with us today!
11349006
View Documents
Thermo Scientific Alfa Aesar
L20178.06
5 g
203.01 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

Solubility
Not miscible or difficult to mix with water.

Notes
Mositure sensitive. Store away from oxidizing agents and water/moisture.
Specifications

Chemical Identifiers

994-89-8
315.088
YEMJHNYABQHWHL-UHFFFAOYSA-N
621176
CCCC[Sn](CCCC)(CCCC)C#C
C14H28Sn
MFCD00009420
tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
tributyl(ethynyl)stannane

Specifications

994-89-8
70°C (0.2 mmHg)
C14H28Sn
1.477
UN2788
Moisture sensitive
Not miscible or difficult to mix with water.
CCCC[Sn](CCCC)(CCCC)C#C
315.088
315.09
Ethynyltri-n-butyltin
1.08
73°C (163°F)
HC≡C-Sn(CH2CH2CH2CH3)3
MFCD00009420
3537659
tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
YEMJHNYABQHWHL-UHFFFAOYSA-N
tributyl(ethynyl)stannane
621176
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H227-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P210-P260-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H301-H312-H315-H319-H360FD-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.