missing translation for 'onlineSavingsMsg'
Learn More
Eugenol, 99%, Thermo Scientific Chemicals Product Code.: 11406897

Eugenol, 99%, Thermo Scientific Chemicals

Product Code. 11406897
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11406897

Brand: Thermo Scientific Alfa Aesar A14332.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Internal standard in GC assays for presence of thymol in biological samplesEugenol is used in cosmetics, essential oil, perfumeries and flavorings. It finds application as a local antiseptic and anesthetic. It is combined with zinc oxide to form zinc oxide eugenol, which is used in dental restoration especially for root canal sealing and prosthodontics. It is also an antifungal antimicrobial agonist of the TRPV1 receptor. It is also used in production of stabilizers and antioxidants for plastics and rubbers. Further, it is used in the production of vanillin. It serves as an active ingredient in cloves. In addition to this, it is used as an insect attractant, chemical intermediate and denaturant for alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Internal standard in GC assays for presence of thymol in biological samplesEugenol is used in cosmetics, essential oil, perfumeries and flavorings. It finds application as a local antiseptic and anesthetic. It is combined with zinc oxide to form zinc oxide eugenol, which is used in dental restoration especially for root canal sealing and prosthodontics. It is also an antifungal antimicrobial agonist of the TRPV1 receptor. It is also used in production of stabilizers and antioxidants for plastics and rubbers. Further, it is used in the production of vanillin. It serves as an active ingredient in cloves. In addition to this, it is used as an insect attractant, chemical intermediate and denaturant for alcohol.

Solubility
Miscible with ethanol, ether, oils, alcohol, glacial acetic acid, chloroform and aqueous sodium hydroxide. Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008654
InChI Key RRAFCDWBNXTKKO-UHFFFAOYSA-N
Synonym eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol
PubChem CID 3314
ChEBI CHEBI:4917
IUPAC Name 2-methoxy-4-prop-2-enylphenol
SMILES COC1=CC(CC=C)=CC=C1O

Specifications

Melting Point -10°C
Density 1.068
Boiling Point 252°C to 253°C
Flash Point 127°C (260°F)
Odor Pungent
Refractive Index 1.541
Quantity 100 g
Beilstein 1366759
Sensitivity Air Sensitive
Merck Index 14,3898
Solubility Information Miscible with ethanol,ether,oils,alcohol,glacial acetic acid,chloroform and aqueous sodium hydroxide. Immiscible with water.
Formula Weight 164.21
Percent Purity 99%
Chemical Name or Material Eugenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242570 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog NumberA14332.36,A14332.22
Lot NumberU17I031 Certificate TypeCertificate of Analysis Date24/08/2022 Catalog NumberA14332.36,A14332.22
Lot Number10240458 Certificate TypeCertificate of Analysis Date05/05/2022 Catalog NumberA14332.36,A14332.22
Lot Number10139077 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14332.36,A14332.22
Lot Number10185463 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14332.36,A14332.22
compliance-icons
Product Identifier
  • Eugenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Eugenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eugenol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.