missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Eukaryotic 18S rRNA Endogenous Control (FAM™/MGB probe, non-primer limited)
GREENER_CHOICE

Eukaryotic 18S rRNA Endogenous Control (FAM™/MGB probe, non-primer limited)

350.00 € - 1208.00 €

Specifications

Accession Number X03205.1
Amplicon Size 187
Content And Storage Control Primer-Probe Set(s)
Detection Method Primer-probe
Format Tube
View More Specs

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10370545
GREENER_CHOICE
missing translation for 'viewDocuments'
Applied Biosystems™
4333760T
125 Reactions
350.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10015904
GREENER_CHOICE
missing translation for 'viewDocuments'
Applied Biosystems™
4333760F
500 Reactions
724.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10618295
GREENER_CHOICE
missing translation for 'viewDocuments'
Applied Biosystems™
4352930E
2500 Reactions
1208.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Applied Biosystems™ Eukaryotic 18S rRNA Endogenous Control (FAM™ MGB Probe, Non-Primer Limited) is intended as an endogenous control. It allows relative gene expression quantification in cDNA samples when used with other gene expression assays. Probe is labeled with 6FAM™ dye - MGB and the primers are not limited. Can be used for singleplex PCR reactions. Endogenous control is to be used with Inventoried and Non-Inventoried TaqMan™ Gene Expression Assays, Custom TaqMan™ Gene Expression Assays, and⁄or Custom TaqMan™ Primers and Probes.

Assay Details:

Gene Symbol: 18S
RefSeq: X03205.1
Probe Exon Location: NA
Amplicon Size: 187
Corresponding TaqMan Assay ID: Hs99999901_s1

TaqMan™ Endogenous Controls

Eliminate months of assay design, formulation, and testing by using TaqMan™ Endogenous Controls as your controls to quantify gene expression. This convenient collection of pre-designed probe and primer sets enables you to normalize the amount of sample RNA or DNA in a reaction.

Complete Solution for Quantitative Gene Expression

Having a hard time deciding what controls to use to quantify gene expression—even with detailed information on biological systems? Now, with TaqMan™ Endogenous Controls, you can avoid all the trial-and-error of selecting controls for most common human, mouse, rat, and eukaryotic genes.

Simple to Use

All components of the TaqMan™ Endogenous Controls are QC tested, formulated into a single 20X mix, and functionally tested. The controls are not only simple to use, but they are also fully compatible with universal conditions for two-step RT-PCR. Just add TaqMan™ Universal PCR Master Mix (with or without AmpErase™ UNG) and your cDNA sample to generate sensitive, reproducible, and truly quantitative gene expression data on Applied Biosystems instruments including the Applied Biosystems 7900HT, 7300, 7500 Real-Time PCR Systems, and the 7000 and 7700 Sequence Detection Systems.

Flexible Offering

We build each endogenous control using our proven 5' nuclease chemistry. For maximum flexibility, you can choose between two different reporter dyes and two quenchers:
  • A FAM™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (900nM each)
  • A VIC™ dye-labeled TaqMan™ MGB probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)
  • A VIC™ dye-labeled TAMRA™ probe (250nM, final concentration) and two unlabeled PCR primers (150nM each y primer limited)

    Choosing the Right Endogenous Control

    Endogenous controls can normalize the expression levels of target genes by correcting differences in the amount of cDNA that is loaded into PCR reaction wells. For best results, verify that the endogenous control is consistently expressed in the sample set to be tested. Endogenous control expression must be uniform across all samples in the study. For multiplexing, ensure that the gene expression level of the endogenous control is greater than that of the target.

    Multiplex vs. Singleplex PCR

    All TaqMan™ Endogenous Controls that contain probes labeled with the VIC™ reporter dye are primer limited. This allows multiplexing of TaqMan™ Endogenous Controls with target gene expression assays, provided that the control gene is more abundantly expressed than the target gene. All TaqMan™ Endogenous Controls that contain probes labeled with the FAM™ reporter dye are not primer limited and are not intended for multiplexing .

    For Research Use Only. Not for use in diagnostic procedures.
  • Specifications

    Specifications

    X03205.1
    Control Primer-Probe Set(s)
    Tube
    18S
    FAM
    Room Temperature
    18S rRNA
    20X
    187
    Primer-probe
    2-step RT-qPCR
    Sustainable packaging
    FAM™, TaqMan™
    Eukaryotic, Rat, Mouse, Human
    Gene Expression
    Videos
    SDS
    Documents

    Documents

    Certificates

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.