missing translation for 'onlineSavingsMsg'
Learn More

Europium foil, 0.25mm (0.01in) thick, 99.9% (REO)

Product Code. 11361228
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
This item is not returnable. View return policy

Product Code. 11361228

Brand: Thermo Scientific Alfa Aesar 010175.FF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-53-1 | Eu | 151.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-53-1
Molecular Formula Eu
Molecular Weight (g/mol) 151.96
MDL Number MFCD00010992
InChI Key OGPBJKLSAFTDLK-UHFFFAOYSA-N
Synonym europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
PubChem CID 23981
ChEBI CHEBI:32999
IUPAC Name europium
SMILES [Eu]

Tekniske data

Color Gray
Quantity 25 x 25 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.82g/25x25mm
Form Foil
UN Number UN3208
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Europium foil, 0.25mm (0.01 in.) thick
compliance-icons
Produktidentifikator
  • Europium foil
Signalord
  • Danger
Farekategori
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 2
Faresætning
  • H250-Catches fire spontaneously if exposed to air.
  • H261-In contact with water releases flammable gases.
Sikkerhedssætning
  • P223-Do not allow contact with water.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P422-Store contents under .
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.