missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium foil, 1.0mm (0.04 in.) thick, 99.9% (REO), Thermo Scientific™
Click to view available options
Quantity:
25 x 25 mm
Packaging:
Mylar
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Packaged under argon
Specifications
Specifications
CAS | 7440-53-1 |
Molecular Formula | Eu |
MDL Number | MFCD00010992 |
UN Number | UN3208 |
Quantity | 25 x 25 mm |
Synonym | europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil |
Solubility Information | Reacts with water. |
InChI Key | OGPBJKLSAFTDLK-UHFFFAOYSA-N |
SMILES | [Eu] |
IUPAC Name | europium |
Show More |
Safety and Handling
Product Identifier
- Europium foil
- Danger
- Pyrophoric solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H250-Catches fire spontaneously if exposed to air.
- H261-In contact with water releases flammable gases.
- P223-Do not allow contact with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P222-Do not allow contact with air.
- P231+P232-Handle under inert gas. Protect from moisture.
- P422-Store contents under .
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu