missing translation for 'onlineSavingsMsg'
Learn More

Europium(II) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11328898
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14077-39-5
Molecular Formula EuF2
Molecular Weight (g/mol) 189.961
MDL Number MFCD01866333
InChI Key YLDGIMHVWSTRML-UHFFFAOYSA-L
Synonym europium ii fluoride, acmc-20akvn, europium ii fluoride, anhydrous
PubChem CID 21225127
IUPAC Name europium(2+);difluoride
SMILES [F-].[F-].[Eu+2]

Specifications

Boiling Point >2400°C
Melting Point 1380°C
Physical Form Powder
Quantity 2 g
UN Number UN3288
Solubility Information Insoluble in water.
Formula Weight 189.96
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.495 g/mL
Assay (metals basis)
Chemical Name or Material Europium(II) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'