missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
145.73 € - 586.92 €
Chemical Identifiers
CAS | 62667-64-5 |
---|---|
Molecular Formula | C6H9EuO6 |
Molecular Weight (g/mol) | 329.10 |
MDL Number | MFCD00150115 |
InChI Key | LNYNHRRKSYMFHF-UHFFFAOYSA-K |
Synonym | europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis |
PubChem CID | 16212055 |
SMILES | [Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360118
|
Thermo Scientific Alfa Aesar
042878.06 |
5 g |
145.73 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370118
|
Thermo Scientific Alfa Aesar
042878.14 |
25 g |
586.92 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Europium(III) acetate hydrate is used as fluorescent powder and as a chemical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62667-64-5 | |
329.10 | |
LNYNHRRKSYMFHF-UHFFFAOYSA-K | |
16212055 |
C6H9EuO6 | |
MFCD00150115 | |
europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis | |
[Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Specifications
62667-64-5 | |
Powder | |
C6H9EuO6 | |
europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis | |
LNYNHRRKSYMFHF-UHFFFAOYSA-K | |
europium(3+) triacetate | |
16212055 | |
Hygroscopic | |
Europium(III) acetate hydrate |
White | |
99.9% (REO) | |
MFCD00150115 | |
Soluble in water. | |
[Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
329.10 | |
329.10 (Anhydrous) | |
White crystalline powder |
Safety and Handling
missing translation for 'einecsNumber' : 214-670-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only