missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) carbonate hydrate, REacton™, 99.99% (REO)

Product Code. p-4408086
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11316668

Brand: Thermo Scientific Alfa Aesar 013212.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86546-99-8 | C3Eu2O9 | 483.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86546-99-8
Molecular Formula C3Eu2O9
Molecular Weight (g/mol) 483.95
MDL Number MFCD00149702
InChI Key MBFNJTXDYDGXDL-UHFFFAOYSA-H
Synonym europium iii carbonate hydrate, 3co3.2eu.h2o, europium iii carbonate hydrate, reacton, dieuropium 3+ hydrate tricarbonate
PubChem CID 91886282
IUPAC Name dieuropium(3+) tricarbonate
SMILES [Eu+3].[Eu+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (REO)
Formula Weight 483.95 (Anhydrous)
Odor Odorless
Appearance White powder
Chemical Name or Material Europium(III) carbonate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.