missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) chloride, ultra dry, 99.99% (REO), Thermo Scientific Chemicals

106.53 € - 1192.94 €

Chemical Identifiers

CAS 10025-76-0
Molecular Formula Cl3Eu
Molecular Weight (g/mol) 258.31
MDL Number MFCD00010994
InChI Key NNMXSTWQJRPBJZ-UHFFFAOYSA-K
Synonym acmc-20akvq, europium 3+ trichloride, europium 3+ ion trichloride, europium iii chloride, ultra dry reo, europium chloride, anhydrous, europium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 1g
PubChem CID 9859933
SMILES [Cl-].[Cl-].[Cl-].[Eu+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388607
SDS View Documents
Thermo Scientific Alfa Aesar
035738.03
1 g
106.53 €
1g
Please to purchase this item. Need a web account? Register with us today!
11398607
SDS View Documents
Thermo Scientific Alfa Aesar
035738.06
5 g
346.73 €
5g
Please to purchase this item. Need a web account? Register with us today!
11455050
SDS View Documents
Thermo Scientific Alfa Aesar
035738.14
25 g
1192.94 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Europium(III) chloride is used as a starting material for the preparation of other europium salts. It is also used in preparation of europium(II)chloride by reduction in a gold boat using hydrogen gas. Further, it is also used to prepare organometallic compounds of europium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10025-76-0
258.31
NNMXSTWQJRPBJZ-UHFFFAOYSA-K
9859933
Cl3Eu
MFCD00010994
acmc-20akvq, europium 3+ trichloride, europium 3+ ion trichloride, europium iii chloride, ultra dry reo, europium chloride, anhydrous, europium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 1g
[Cl-].[Cl-].[Cl-].[Eu+3]

Specifications

850°C
Powder
MFCD00010994
acmc-20akvq, europium 3+ trichloride, europium 3+ ion trichloride, europium iii chloride, ultra dry reo, europium chloride, anhydrous, europium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 1g
NNMXSTWQJRPBJZ-UHFFFAOYSA-K
europium(3+);trichloride
9859933
99.99%
Hygroscopic
Europium(III) chloride, Ultra dry
10025-76-0
Cl3Eu
14,3904
Soluble in water; Soluble in dilute hydrochloric acid,acetone and alcohol; Slightly soluble in tetrahydrofuran.
[Cl-].[Cl-].[Cl-].[Eu+3]
258.31
258.32
Ampule
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 233-040-4

missing translation for 'rtecsNumber' : LE7525000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.