missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Chemical Identifiers

CAS 13765-25-8
Molecular Formula EuF3
Molecular Weight (g/mol) 208.96
MDL Number MFCD00010998
InChI Key HPNURIVGONRLQI-UHFFFAOYSA-K
Synonym europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
IUPAC Name europium(3+) trifluoride
SMILES [F-].[F-].[F-].[Eu+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336918
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013659.04
2 g N/A
N/A
11326918
View Documents Promotion Details
Thermo Scientific Alfa Aesar
013659.09
10 g N/A
N/A
Description

Description

Europium(III) fluoride is used as a catalyst in the preparation of novel mixed-metal fluorides. It finds application in studies related to fluoride glasses. It is also used as a laboratory reagent. Further, it is used in optical glasses and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13765-25-8
208.96
HPNURIVGONRLQI-UHFFFAOYSA-K
europium(3+) trifluoride
EuF3
MFCD00010998
europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
[F-].[F-].[F-].[Eu+3]

Specifications

>2280°C
13765-25-8
EuF3
europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
HPNURIVGONRLQI-UHFFFAOYSA-K
europium(3+) trifluoride
208.96
Reacton™
(REO)
1390°C
Crystalline
MFCD00010998
Slightly soluble in strong mineral acids. Insoluble in water.
[F-].[F-].[F-].[Eu+3]
208.96
99.9%
Hygroscopic
Europium(III) fluoride, Anhydrous
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 237-368-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only