missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) nitrate hexahydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11387108
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Europium(III) nitrate hexahydrate is used in the preparation of pharmaceuticals, agrochemicals and other chemical products. It is used as a catalyst and a component of specialized steels. It is also used in the synthesis of light emitting nanomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-53-5
Molecular Formula EuH12N3O15
Molecular Weight (g/mol) 446.066
MDL Number MFCD00149700
InChI Key JVYYYCWKSSSCEI-UHFFFAOYSA-N
Synonym europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1g
PubChem CID 202256
IUPAC Name europium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3]

Specifications

Melting Point 85°C
Color Colorless
Physical Form Crystalline or Powder
Quantity 2 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 446.06 (337.97 Anhydrous)
Odor Odorless
Appearance Colorless crystalline
Chemical Name or Material Europium(III) nitrate hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'