missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Europium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Europium(III) nitrate hydrate is widely used as dopant and used in the preparation of europium-doped hafnium and europium doped gadolinium oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 85°C |
Color | Colorless to Pink |
Physical Form | Crystalline Aggregates |
Quantity | 5 g |
UN Number | UN1477 |
Merck Index | 14,3904 |
Solubility Information | Highly soluble in water. |
IUPAC Name | europium(3+);trinitrate;hydrate |
Formula Weight | 337.97 (Anhydrous) |
Percent Purity | 99.99% |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction