missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) nitrate hydrate, REacton™, 99.99% (REO)

Product Code. p-7048707
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11371029

Brand: Thermo Scientific Alfa Aesar 011296.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100587-95-9 | EuN3O9 | 337.98 g/mol

Europium(III) nitrate hydrate is widely used as dopant and used in the preparation of europium-doped hafnium and europium doped gadolinium oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100587-95-9
Molecular Formula EuN3O9
Molecular Weight (g/mol) 337.98
MDL Number MFCD00149700
InChI Key GAGGCOKRLXYWIV-UHFFFAOYSA-N
Synonym europium iii nitrate hydrate, acmc-1c6y6, eu.3no3.h2o, europium trinitrate monohydrate, europium 3+ hydrate trinitrate, europium 3+ ion hydrate trinitrate, nitric acid,europium 3+ salt, hydrate 9ci
PubChem CID 57346089
SMILES [Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 85°C
Color Colorless to Pink
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1477
Merck Index 14,3904
Solubility Information Highly soluble in water.
IUPAC Name europium(3+);trinitrate;hydrate
Formula Weight 337.97 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Assay (REO)
Chemical Name or Material Europium(III) nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.