missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxide, nanopowder, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11479860
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Europium oxide nanopowder acts as phosphors for lamps. It is used as a fluorescent label in immunoassay for atrazine. It is also used in color TV, X-ray, glass, crystal and other luminescent materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
PubChem CID 159371
IUPAC Name dieuropium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Melting Point 2291°C
Physical Form Powder
Quantity 5 g
Merck Index 14,3904
Solubility Information Insoluble in water.
Formula Weight 351.92
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium (III) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'