Learn More
Europium(III) oxide, REacton™, 99.9% (REO)
Beskrivelse
Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tekniske data
Tekniske data
| Melting Point | 2291°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.9% (REO) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 351.92 |
| Sensitivity | Hygroscopic |
| Appearance | White powder or chunks |
| Density | 7.42g/cm3 |
| Vis mere |
RUO – Research Use Only
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.