missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxide, REacton™, 99.99% (REO)

Product Code. 11351039
undefined
Quantity:
1 g
5 g
25 g
100 g
missing translation for 'unitSize'
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11351039

missing translation for 'mfr': Thermo Scientific Alfa Aesar 011299.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol

Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
PubChem CID 159371
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Melting Point 2291°C
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water.
IUPAC Name europium(3+);oxygen(2-)
Formula Weight 351.92
Sensitivity Hygroscopic
Appearance White powder or chunks
Chemical Name or Material Europium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.