missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific™

Product Code. 11400301
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13537-22-9
Molecular Formula Cl3EuO12
Molecular Weight (g/mol) 450.30
MDL Number MFCD00016076
InChI Key KDBBDPJZIDRZFY-UHFFFAOYSA-K
Synonym europium perchlorate, europium 3+ perchlorate, acmc-1aedp, europium iii perchlorate solution, trihyperchloric acid europium iii salt, europium iii perchlorate aqueous sol.
PubChem CID 11733073
IUPAC Name europium(3+);triperchlorate
SMILES [Eu+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 2 g
UN Number UN3211
Formula Weight 558.40 (450.32 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Europium(III) perchlorate hexahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.