missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) sulfate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

74.17 € - 297.48 €

Chemical Identifiers

CAS 10031-55-7
Molecular Formula Eu2H16O20S3
Molecular Weight (g/mol) 736.216
MDL Number MFCD00149704
InChI Key PGDXJFDXKJLVTJ-UHFFFAOYSA-H
Synonym europium iii sulfate octahydrate, 2eu.3so4.8h2o, europium sulfate, europium sulfate,, dieuropium 3+ octahydrate trisulfate, europium 3+ sulfate-water 2/3/8
PubChem CID 25021676
IUPAC Name europium(3+);trisulfate;octahydrate
SMILES O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11302668
SDS View Documents
Thermo Scientific Alfa Aesar
029106.04
2 g
74.17 €
2g
Please to purchase this item. Need a web account? Register with us today!
11392658
SDS View Documents
Thermo Scientific Alfa Aesar
029106.09
10 g
297.48 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Europium(III) sulfate octahydrate is used as a phosphor activator for color cathode-ray tubes and liquid-crystal displays. It is used in computer monitors and televisions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10031-55-7
736.216
PGDXJFDXKJLVTJ-UHFFFAOYSA-H
25021676
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3]
Eu2H16O20S3
MFCD00149704
europium iii sulfate octahydrate, 2eu.3so4.8h2o, europium sulfate, europium sulfate,, dieuropium 3+ octahydrate trisulfate, europium 3+ sulfate-water 2/3/8
europium(3+);trisulfate;octahydrate

Specifications

375°C (in -8H2O)
Crystalline
MFCD00149704
europium iii sulfate octahydrate, 2eu.3so4.8h2o, europium sulfate, europium sulfate,, dieuropium 3+ octahydrate trisulfate, europium 3+ sulfate-water 2/3/8
PGDXJFDXKJLVTJ-UHFFFAOYSA-H
europium(3+);trisulfate;octahydrate
25021676
99.9%
Hygroscopic
Europium(III) sulfate octahydrate
10031-55-7
Eu2H16O20S3
14,3904
Soluble in water.
O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Eu+3].[Eu+3]
736.216
736.23 (592.10 Anhydrous)
Odorless
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 236-903-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.