missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) trifluoroacetate hydrate, 99.9% (metals basis), Thermo Scientific™

Product Code. 11475450
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Eu(OOCCF3)3·xH2O
MDL Number MFCD05663782

Specifications

Physical Form Crystalline
Quantity 10 g
UN Number UN3288
Formula Weight 490.97 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Europium (III) trifluoroacetate hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'