missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Product Code. 15491647
Click to view available options
Quantity:
10 g

 

It is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52093-25-1
Color White
Physical Form Powder
Quantity 10 g
Molecular Formula C3EuF9O9S3
Linear Formula Eu(OSO2CF3)3
MDL Number MFCD00209587
Synonym europium trifluoromethanesulfonate, europium iii trifluoromethanesulfonate, europium iii triflate, europium triflate, eu otf 3, ksc268k4f, europium 3+ tritriflate, europium iii tris trifluoromethanesulfonate, europium 3+ ion tritrifluoromethanesulfonate
Solubility Information Insoluble in water.
InChI Key TWNOVENTEPVGEJ-UHFFFAOYSA-K
SMILES [Eu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
IUPAC Name europium(3+);trifluoromethanesulfonate
Molecular Weight (g/mol) 599.15
PubChem CID 9960317
Formula Weight 599.15
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Europium(III) trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'