missing translation for 'onlineSavingsMsg'
Learn More

IKA™ EUROSTAR 100 Overhead Stirrers
GREENER_CHOICE

Use this powerful laboratory mixer for a variety of applications. IKA™ EUROSTAR 100 Overhead Stirrer automatically adjusts the speed through microprocessor controlled technology.

Brand:  IKA™ 0004238102

1534.96 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 13591900

  • 2900.00 € / Each
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Equipped with a RS 232 and a USB interface to control and document all parameters
  • Integrated torque trend display is provided for the measurement of viscosity changes
  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • Guarantees constant speed even with changes in viscosities of the sample
  • Multilingual TFT display
  • Programmable functions
  • Integrated temperature measurement
  • Interval operation
  • Reverse function
  • Timer function
  • Adjustable safety circuit
  • Locked function
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Quiet operation
  • Error code Display
  • H 67.60 temperature sensor and WH 11 WiCo holder included in delivery
Specifications

Specifications

30 to 1300 rpm
7 Segment LED
186 w
248 mm
230V, 50/60Hz, 186w
Alu-Cast Coating/Thermoplastic Polymer
5°C to 40°C
70,000 mPas
100 L
230 V
H x W x dia.: 248 x 86 x 208 mm
86 mm
50/60 Hz
EUROSTAR 100 Digital
100 Ncm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers