missing translation for 'onlineSavingsMsg'
Learn More

IKA™ EUROSTAR 20 Digital with UK Plug
GREENER_CHOICE

Laboratory stirrer designed for simple stirring tasks for quantities up to 15 L (H2O).

Brand:  IKA™ 0004442002

1093.66 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 16238201

  • 1246.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions
  • Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically
  • This guarantees a constant speed even with changes in viscosities of the sample
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
Specifications

Specifications

30 to 2000 rpm
LED
70 w
50/60 Hz
4.4 kg
15 L
100/230 V
208 x 86 x 248 mm
5°C to 40°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.