Learn More
Thermo Scientific™ Eutech™ 10.0 NTU Calibration Standard for TB1000
The CyberScan TB1000 series turbidity meters and its accessories and standards are designed to meet the stringent requirements of today's scientists.
Brand: Thermo Scientific™ 01X344318
Beschreibung
Eutech™ 10.0 NTU Calibration Standard is used to calibrate the turbidimeter Cyberscan TB1000. The CyberScan TB1000 series turbidity meters and its accessories and standards are designed to meet the stringent requirements of today's scientists.
Features:
- 125mL bottle
- Used With: CyberScan TB1000

Spezifikation
Caibration Standard | |
CyberScan TB 1000 | |
10.0 NTU Calibration Standard | |
125 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.