Learn More
Thermo Scientific™ Eutech TN-100 Handheld Infrared Turbidity Meter Kit
Waterproof, portable, infrared Turbidity meter wtih extended range up to 2000 NTU, 0.01 resolution
Brand:  Thermo Scientific™ 01X357301
920.38 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 10142422
  • 968.82 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Includes: Set of 4 calibration standards (800, 100, 20.0 & 0.02 NTU)
Description
Description
Waterproof, portable, infrared Turbidity meter wtih extended range up to 2000 NTU, 0.01 resolution
  • Waterproof
  • Set of 7 sample vials
  • Lint-free cloth
  • Silicon oil
  • Batteries
  • Hard carrying case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Infrared Turbidity Meter Kit
U.S. EPA-approved non-formazin standards
0.01
ISO 7027 compliant
Portable, Waterproof
Waterproof TN-100 handheld turbidimeter with set of 4 calibration standards (800, 100, 20.0 & 0.02 NTU), set of 7 sample vials, lint-free cloth, silicone oil, batteries, and hard carrying case.
Up to 2000 NTU
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Eutech TN-100 Handheld Infrared Turbidity Meter Kit >