Learn More
Thermo Scientific™ Eutech™ ATC Electrode for pH 5+ Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. 
Brand:  Thermo Scientific™ PH5TEM01P
Product Code. 10152612
  • 72.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH 5+ pH Meter with single Junction pH electrode is an ideal choice for routine applications in laboratories, productions plants and schools. The ATC (Automatic Temeprature Compensation) electrode ECPH5TEM01P comes with the Eutech™ pH 5+ meter.
 Used with:
  • ph 5+meter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATC Electrode for pH 5+ Meter
Large display screen
Anti-roll Design, Dual Range