Learn More
Thermo Scientific™ Eutech™ Automatic Temperature Compensation (ATC) Probe ECPHWPTEM03J
Eutech™ offers a wide range of temperature probes used in various educational, laboratory, and industrial applications. 
Brand:  Thermo Scientific™ 01X021820
Product Code. 12971688
  • 118.59 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Eutech™ offers a wide range of temperature probes used in various educational, laboratory, and industrial applications. Eutech™ Automatic Temperature Compensation (ATC) Probe ECPHWPTEM03J is used with the ph 600 series meters.
Features:
  • 3 meter cable length
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATC Probe
Eutech™ pH 620, 610, 600, PC650, PD650, PCD650 meters
Probe/Electrode
3 m