Learn More
Eutech™ Combination pH/Conductivity/ATC Probe COMBI03BW for Eutech™ PC 450
Optimize your Eutech™ conductivity meters with the combination pH/Conductivity/ATC probes.
Brand: Thermo Scientific Eutech 01X230205
181.53 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
The Eutech™ Combination pH/Conductivity/ATC Probe COMBI03BW is used with the Eutech™ CON 450 Conductivity Meter. The Eutech™ CON 450 Conductivity Meter is a backlit, rugged waterproof meter ideal for any environment. Durable waterproof and dustproof design with IP67 rating. Large, backlit custom display so you can view measurement with temperature and cell efficiency.
Features:- 3 meter cable
- For Use with: Eutech PC 450
Specifications
pH/Conductivity/ATC Combination Probe |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.