missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ CyberScan PC 650 Multi-Parameter Meter
Click to view available options
Measurement Modes:
pH, mV, Ion, Conductivity, TDS, Resistivity, Salinity
Unit Size:
Each
Description
Combining two of the most popular electrochemistry parameters, the Eutech’s CyberScan PC650 allows you to measure pH, conductivity and temperature with one handheld, at the same time. For use in industrial, environmental, water quality testing, an aquaculture applications.
Features:
- Simultaneouslymeasures and displays three parameters
- ±1% full scale accuracy at 3-decimal resolution across a widemeasurement range
- Ion-measurement capabilities with 3-digit resolution
- Accepts two-cell and four-cell conductivity probe tomeasure up to 500m/s —metermeasures pure water
- Selectablemulti-point calibration formore accurate conductivitymeasurements or single-point calibration for quick, convenient calibration with nomistakes
- One-Glance-Tells-All Screen Display with Backlight
- Electrode status report, calibration data and simultaneous display of up to 4 parameters
- Rugged and IP67-rated waterproof for applications in harsh environments.meter floats on water
- Easy-to-understand promptmessages for easy set-up, calibration and trouble-shooting
- Recorded readings individually stamped with time and date in GLP compliant format
- Auto-loggingmeter saves readings automatically within seconds to prevent data loss
- HI-LO limit alarm for easy quality control –meter alerts when readings fall outside selected ranges
- Wireless Information Technology: No wires, no cables—Transfer data frommeter to computer with the press of a button
- Non-Volatilememory protects information andmeter settings, even when batteries run out
- Four Levels of Password Protection prevents tampering or accidental data deletion
- Calibration Alarm: Nomore out-dated calibration settings
Specifications:
- pH Range: -2.000 to 20.000 pH
- No. of pH Calibration Points:1 (Offset) to 6 points
- pH Buffer OptionUSA, NIST, DIN, PWB, Custom
- mV Range: ±2000.0mV
- Ion Concentration Range: 0.001 to 19900
- No. of Ion Calibration Pts : Up to 8
- Conductivity Range:.. to 500mS/cm
- Cell Constant: 0.010 to 10.000
- Temperature Coefficient: Linear & Pure
- TDS Range: .. to 500.0 ppt
- Salinity Range: 0 to 80 ppt
- Resolution & Accuracy: 0.05% FS & ±1% FS +1 LSD
- Temperature Range: -10.0 to 110.0 °C / 14.0 to 230.0 °F (meter)
- Temperature Compensation: Automatic /manual (0 to 100 °C)
- memory: 500 Data Sets with Date/Time stamping
- Security: password protection
- Output: IrDA Wireless Communications
- Display:multi-data Back-lit dotmatrix; 7.2 cm (H) X 5.4 cm (W)
- Special Functions: Auto-Calibration; Data-logging options with time/date indexed, GLP compliance; Simultaneous Display andmeasurement of 4 parameters; Cal-due alarm; pH slope/offset Display & Self-diagnostic
- Power: 4 X 1.5 V 'AA' Alkaline batteries or 9 V DC adapter, 500mA
Specifications
Specifications
| Outputs | IrDA Wireless Communication |
| Description | Waterproof CyberScan PC 650 pH/ mV/ Ion/ Conductivity/ TDS/ Resistivity/ Salinity handheld meter with pH electrode ECFC252203B, 4-cell Conductivity probe, DAS software, power adapter & carrying kit set (all probes comes with 3m long cable.) |
| Display Type | LCD Graphic, Backlit |
| IP Rating | IP67 |
| Measurement Modes | pH, mV, Ion, Conductivity, TDS, Resistivity, Salinity |
| Range (pH) | -2 to 20 |
| Range (Salinity) | 0 to 80 ppt |
| Temperature Range (Metric) | -10°C to +110°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction