missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ CyberScan pH 610 pH Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Brand: Thermo Scientific™ 01X415106
759.12 EUR valid until 2024-12-31
Use promo code "21937" to get your promotional price.
This item is not returnable.
View return policy
Description
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ CyberScan pH 610 pH Meter is an ideal choice for routine applications in laboratories, productions plants and schools.
Features:
- 6 calibrations points and 20 auto/manual buffer options sets for quick, convenient calibration.
- Cal-Due Alarm Prevents Out-dated Calibrations, while Set-Point Alarms warns when readings are outside set-point limits.
- Higher accuracy under varying temperature with Automatic/manual Temperature Compensation.
- mega-memory space that stores/ recalls up to 500 Data Sets with temperature readings, with time and date-stamping formatted tomeet GLP standards.
- Wireless and cable-free data transfer with infrared IrDA technology. Comes with PC-based Complimentary Data Acquisition Software for hassle-free data import and report-generation into any popular spreadsheet programs of your choice.
- Recall Electrode Slope, Electrode Offset Value and Previous Calibration Values for easy reference
- Large, Easy-to-Read Custom Display with simultaneous readings provides optimum viewing with user-friendly icons
- Hold Functionmomentarily freezes reading for easy viewing and recording
- Self-Diagnostic withmessage codes for easy troubleshooting
- Volatilememory holds settings and stored data even when you run out of batteries.
- Rugged and waterproof to IP67 standard with protective rubber boot and handy velcro strap for better grip.
- Power with batteries or optional AC adapter for use in the field or at bench-top with built-in stand.
- Energy-saving with Auto-power off feature.
Specifications:
- pH Range: -2.000 to 19.999 pH
- mV Range: ±2000mV
- Temperature Range: -10.0 to 110.0 °C (14.0 to 230 °F)
- Temperature Compensation: Automatic /manual (0 to 100 °C)
- No. of Calibration Points: Up to 6 points
- Cal Due Alarm: Yes (Days -max 30)
- pH Buffer Options: USA, NIST, DIN, PWB
- Custom Buffer Calibration: Yes (2 to 5)
- Slope/ Offset Display: Yes (Display + Icon)
- Inputs: DC phono sockets, 8 pin connector, BNC
- LCD Display: Back-lit Dotmatrix
- memory: 500 data sets
- Output: Yes (IrDA/ RS232C)
- Special Functions: Stability Indicator; Self-diagnostic; Non-Volatilememory; Hold & Auto-Power Off after 20minutes
- GLP: Yes
- Ingress Protection: IP67
- Power: 4 'AAA' x 1.4V batteries or 9VDC power adapter
Specifications
IrDA, RS-232C | |
-10°C to +110°C | |
Yes |
Up to 6 | |
LCD | |
500 |