missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ CyberScan pH 620 pH Meter

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. 

Brand:  Thermo Scientific™ 01X415107

827.30 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 10309412

  • 1356.75 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ CyberScan pH 620 pH Meter is an ideal choice for routine applications in laboratories, productions plants and schools.

Features:

  • measures pH and Ion with up to 0.001 Resolution.
  • Cal-Due Alarm Prevents Out-dated Calibrations, while Set-Point Alarms warns when readings are outside set-point limits.
  • Higher accuracy under varying temperature with Automatic/manual Temperature Compensation.
  • mega-memory space that stores/ recalls up to 500 Data Sets with temperature readings, with time and date-stamping formatted tomeet GLP standards.
  • Wireless and cable-free data transfer with infrared IrDA technology. Comes with PC-based Complimentary Data Acquisition Software for hassle-free data import and report-generation into any popular spreadsheet programs of your choice.
  • Recall Electrode Slope, Electrode Offset Value and Previous Calibration Values for easy reference
  • Large, Easy-to-Read Custom Display with simultaneous readings provides optimum viewing with user-friendly icons
  • Hold Functionmomentarily freezes reading for easy viewing and recording
  • Self-Diagnostic withmessage codes for easy troubleshooting
  • Volatilememory holds settings and stored data even when you run out of batteries.
  • Rugged and waterproof to IP67 standard with protective rubber boot and handy velcro strap for better grip.
  • Power with batteries or optional AC adapter for use in the field or at bench-top with built-in stand.
  • Energy-saving with Auto-power off feature.

Specifications

  • pH Range: -2.000 to 19.999 pH
  • mV Range: ±2000mV
  • Ion Range: 0.001 to 19900
  • Units: ppm/molar/mgL
  • Resolution & Accuracy: 2/ 3 digits & 0.5% FS (monovalent) / 1% (Divalent)
  • Temperature Range: -10.0 to 110.0 °C (14.0 to 230 °F)
  • Temperature Compensation: Automatic /manual (0 to 100 °C)
  • No. of Calibration Points: Up to 6 points
  • Cal Due Alarm: Yes (Days -max 30)
  • pH Buffer Options: USA, NIST, DIN, PWB
  • Custom Buffer Calibration: Yes (2 to 5)
  • Slope/ Offset Display: Yes (Display + Icon)
  • Inputs: DC phono sockets, 8 pin connector, BNC
  • LCD Display: Back-lit Dotmatrix
  • memory: 500 data sets
  • Output: Yes (IrDA/ RS232C)
  • Special Functions: Stability Indicator; Self-diagnostic; Non-Volatilememory; Hold & Auto-Power Off after 20minutes
  • GLP: Yes
  • Ingress Protection: IP67
  • Power: 4 'AAA' x 1.4V batteries or 9VDC power adapter
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IrDA, RS-232C
-10°C to +110°C
Yes
Up to 6
LCD
500
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers