missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eutech Handheld Meters Calibration Solution,500 μS/cm KCl, Thermo Scientific™
Periodic calibration with accurate standards is recommended for meters
Brand: Thermo Scientific Eutech 01X211219
21.27 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- 500 μS/cm KCl calibration solution
- Solutions packed in 480mL durable plastic bottles
- Economical and convenient for laboratory use.
Specifications
Inorganic Salts | |
Odorless | |
102.4°C | |
Eutech Handheld Meters |
7447-40-7 | |
7.0 | |
1 | |
Conductivity Calibration Solution |