missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech Handheld Meter Calibration Solution, 480 mL 111.8 mS/cm KCl
Periodic calibration with accurate standards is recommended for meter
Brand: Thermo Scientific™ 01X211244
28.60 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
Periodic calibration with accurate standards is recommended for meter
- 480 mL 111.8 mS/cm KC calibration solution
- Solutions packed in 480mL durable plastic bottles
- Economical and convenient for laboratory use.
Specifications
Inorganic Salts | |
7447-40-7 | |
Odorless | |
7.0 | |
1 |
111.8 mS / cm KCl | |
Eutech Handheld Meters | |
1.0 g/cm³ at 25°C | |
102.4°C | |
Liquid |