missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 150 Meter Kit

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. 

Brand:  Thermo Scientific™ 01X651607

292.60 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15377043

  • 430.14 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Eutech™ offers a wide range of pH meters and electrodes. pH 150 Meter Kit is an an IP67-rated waterproof pH meter that offers measurement of pH / mV / Temperature.

Features

  • Large, three line LCD – View pH or mV measurement with temperature and % slope
  • Built-in memory function – Stores up to 150 measurements along with temperature
  • Auto buffer recognition – Automatically identifies USA, NIST or DIN buffer sets for rapid calibration
  • Power with AA batteries or use optional AC adapter for benchtop measurements
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
  • Selectable ºF/ºC – Choose units to best fit your application
  • Features mV offset – Calibrate to a Redox/ORP standard or use for pH electrode diagnostics
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • % slope and temperature displayed with each measurement
  • Built-in stand – Ideal for easy benchtop or long-term use
  • Wall-mount option – Ideal to save on bench space
  • Includes innovative Grip-Clip™ – Quickly and easily secures the electrode to your beaker or container
  • Compatible with pH electrodes having a BNC connector
TRUSTED_SUSTAINABILITY
Specifications

Specifications

pH 150 Meter Only: Meter, Grip-Clip™, two AA batteries
±0.2 or ±0.05%, whichever is greater
mV: ±200 mV, Temp: Offset in 0.1 increments, ±10°C or ±18°F
0.01
Up to 5 (USA & NIST), Up to 6 (DIN)
14.0°F to 230.0°F
LCD, 3-Line Custom
150
0.5 kg
Two AA batteries (included), >700 hours continuous use; AC adapter (optional)
Automatic/Manual (-10°C to 110°C)
±0.5°C;/±0.9°F
±0.01
-2 to 16
0.1 mV ±999.9 mV; 1 mV beyond
-10°C to +110°C
0.1°C/°F
Stores up to 150 data sets
7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm)
1 lb.
±2000
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers