Learn More
Thermo Scientific™ Eutech™ pH 5+ pH meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
Brand: Thermo Scientific™ 01X244912
308.20 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH 5+ pH Meter with single Junction pH electrode is an ideal choice for routine applications in laboratories, productions plants and schools.
Features:
- multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
- Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
- Hold Functionmomentarily freezes reading for easy viewing and recording
- Auto-Power Off saves battery power after non-use
- Self-Diagnostic withmessage codes for easy troubleshooting
- Large Custom LCD provides optimum viewing even at a distance
- Protective Rubber Boot shieldsmeter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
- Rugged Carrying Case (Optional) provides handy on-sitemeter calibration andmeasurement
Specifications
- pH Range: 0.00 to 14.00 pH
- Temperature Range: 0 to 100.0 °C
- Temperature Compensation: Automatic /manual (0 to 100 °C)
- No. of Calibration Points: 2 to 5 points
- pH Buffer Options:1.68, 4.01, 7.00, 10.01, 12.45 (USA), 1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
- Special Functions: Auto Power-Off after 17minutes;
- Hold & Self-diagnosticmessages
- Power: 4 'AAA' x 1.4V batteries

Specifications
Eutech pH 5+ pH Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised (Rinse) Water (60 ml Each) | |
2 to 5 | |
0°C to 100°C | |
LCD |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.