Learn More
Thermo Scientific™ Eutech™ pH 700 Meter
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools.
599.00 € - 807.00 €
Specifications
Accuracy (pH) | ±0.01 |
---|---|
Resolution (pH) | 0.01 |
Calibration Points | Up to 5 |
Temperature Range (Metric) | 0°C to 100°C |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11593845
|
Thermo Scientific™
01X541601 |
Eutech pH 700 Meter with 100/240 VAC Adapter | N/A | ||||||
11513855
|
Thermo Scientific™
01X541605 |
Eutech pH 700 Meter With pH Electrode (ECFC7252101B), ATC Probe (PH5TEMB01P), Integral Electrode Holder and 100/240 VAC Adapter | N/A | ||||||
12943517
|
Thermo Scientific™
01X541610 |
Eutech pH 700 Meter With Glass Body, Open Pore pH Electrode (EC620131), ATC Probe (PH5TEMB01P) Integral Electrode Holder & 100/240 VAC Adapter | N/A | ||||||
Description
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools. It features a large, easy-to-read display, ready indicator that letsyou know when readings are stable and up to 5-point calibration with auto-buffer recognition. It also has a non-volatile memory holds up to 100 data points and comes witu an integral electrode holder.
Applications:
- Educational: Useful for most laboratory, ecological studies and other applications.
- Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing. Use in all types of food processing.
Buffer Sets
Specifications
±0.01 | |
0.01 | |
Up to 5 | |
0°C to 100°C |