Learn More
Eutech™ pH Buffers and Solutions
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes
Brand: Thermo Scientific Eutech 01X211203
Description
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.

Specifications
Buffer Solution | |
10.01 | |
Buffer |
pH Meter and Electrode | |
480 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
524-09 | Certificate of Analysis | 27/12/2024 | 01X211203 |
524-04 | Certificate of Analysis | 27/12/2024 | 01X211203 |
524-08 | Certificate of Analysis | 27/12/2024 | 01X211203 |
524-07 | Certificate of Analysis | 27/12/2024 | 01X211203 |
524-06 | Certificate of Analysis | 27/12/2024 | 01X211203 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.