Learn More
Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions
Eutech™ offers Salinity Standard Solutions for use with Salinity Meters.
12.30 € - 16.88 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16445339
|
Thermo Scientific™
01X211231 |
25 ppt NaCl Calibration Solution | N/A | ||||||
15601717
|
Thermo Scientific™
01X211230 |
5 ppt NaCl Calibration Solution |
16.88 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Eutech™ offers a range of Salinity Standard Solutions for use with Salinity Meters. Salinity measurements are common in industries like agriculture, aquaculture, hydroponics, food, pools and spas where it is necessary to monitor the salt level constantly.
Features:
- Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Description:
- ECNACL5PPT: 5 ppt (parts per thousand) Nacl calibration solution
- ECNACL25PPT: 25 ppt (parts per thousand) Nacl calibration solution
- ECNACL45PPT: 45 ppt(parts per thousand) Nacl calibration solution