missing translation for 'onlineSavingsMsg'
Learn More

Eutech™ Salt 6+ Handheld Meters

Get quick and accurate salinity measurements in the field, laboratories, and schools.

Brand:  Eutech™ 01X289429

326.12 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11509214

  • 518.58 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The new Thermo Scientific™ Eutech 6+ series of handheld meters for salinity is ideal for your basic water testing needs.

Innovative electrode design
  • Fast automatic temperature compensation for quick, repeatable measurements, strategically located vents that eliminate air bubble entrapment 
  • Removable electrode sleeve which allows for easy maintenance 
Durable rubber armor with stand
  • Rubber armor with convenient benchtop stand to provide ideal display angle when needed
Impressive salinity accuracy
  • Designed to match the non-linear salinity curve for improved accuracy over the entire range with a single calibration point
Innovative GripClip electrode holder
  • Secures to most beakers without the need for a separate electrode arm
  • Keeps electrodes suspended during use, away from stir bars
  • Allows for quick height adjustment of your electrodes to suite your sample container and individual preference
 Sturdy carrying case with accessories
  • Compact hard case with everything needed for calibration and measurement
  • Protects the meter when not in use and includes a door with room to secure items until needed
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Salt 6+ Meter with Kit, GripClip, Carrying Case
Automatic or Manual (0 to 50°C)
0.1 ppt/0.01%
0.00°C to 100.0°C (80.0°C with Supplied Probe)
±0.5°C (±0.9°F)
>500 hr.
Salinity with Temperature
1.0 to 50.00 ppt/0.1 to 5.00%
±% Full-scale
0.1°C (0.1°F)
4 x 1.5V AAA Alkaline (included)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Warranty: Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.