missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ SMPS Power Adapter for Cyberscan 1500 series meters
The Eutech™ SMPS Power Adapter provides you with the power you need.
Brand: Thermo Scientific™ 60X030131
62.30 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
The Eutech™ SMPS Power Adapter provides you with the power you need. The 100 /240 VAC Power Adapter has US/UK/EUR/Japan plug used for CyberScan 1500 series meters.
Features:
- 12 V, 6 W
- 100/240 VAC
- Used With: CyberScan 1500 meters
Specifications
SMPS Power Adapter. 100/240VAC , with US/UK/EUR/Japan plug , 12V, 6W, CENTRE -ve | |
12 V | |
Cyberscan 1500 series |
SMPS Power Adapter | |
US/UK/EUR/Japan plug |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction