missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Temp 10 Series Temperature Meter

Eutech™ offers a selection of temperature meters for use in various educational, laboratory and industrial applications. 

Brand:  Thermo Scientific™ 01X450408

Product Code. 15398397

  • 234.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

Rugged and easy to use, the Eutech™ Temp 10 Series features a large, backlit dual-line LCD, giving you clear, accurate readings of measured and min/max temperature, even in dark environments. Ergonomically designed to fit your palm perfectly, the Temp 10 series offers maximum hands-free convenience with our exclusive three-way hands-free kit, sold separately.

 Features:

User-Friendly

  • Automatic calibration – just leave electrode in ice water and press ‘CAL’ to calibrate
  • min/max temperature at the press of a key
  • Dual-display shows current andmin/max reading
  • Large, easy-to-read illuminated display

Versatile

  • measures from -250 to 1372 °C
  • Accepts a wide variety of temperature probes using standardmini-connector
  • Exclusive three-way hands-free option* – hang themeter from a pipe or belt, stick themeter to ametallic surface, or simply stand themeter on a table surface

Sturdy Design

  • IP54 splashproof with sealed keypad and ABS plastic housing
  • Protective rubber armor ensures a firm grip and doubles up as a table-top stand
NB: A temperature sensor is not included and must be ordered separately.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dimensions & Weight 175 x 97 x 42mm (LxWxT), 267 g (without armor); 180 x 102 x 52mm (LxWxT), 362 g (with armor)
LCD: 58 x 40mm with backlight
-250°C to +1372°C
-40°C to +65°C (-40°F to +149°F); 10% to 90% (non-condensing)
3 ‘AA’ 1.5 V Alkaline
Min/Max Reading, Auto Shut-Off: 17.5min (selectable), Hold Function,Stability Indicator, Single Input, Ingress Protection: -529 IP54 for dust and water-resistant enclosures
Temperature Meter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers