missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ pH 6+ pH Meter
Click to view available options
Type:
Meter with 3-in-1 Combi Electrode
Meter with pH Electrode
missing translation for 'unitSize'
Each
Description
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH 6+ pH Meter ECPH602PLUSK with single Junction pH electrode and ECPH603PLUSK with 3-in-1 pH/Temperature Combi Electrode are ideal choices for routine applications in laboratories, productions plants and schools.
Features:
- Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
- Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
- Hold Function momentarily freezes reading for easy viewing and recording
- Auto-Power Off saves battery power after non-use
- Self-Diagnostic with message codes for easy troubleshooting
- Large Custom LCD provides optimum viewing even at a distance
- Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
- Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
- pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA),1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
Specifications
Specifications
| Description | Eutech pH 6+ pH Meter with 3-in-1 pH/Temperature Combi Electrode |
| Includes | Eutech pH 6+ pH Meter with 3-in-1 pH/ Temperature Combi Electrode (ECFC7352901B) Carrying Case, 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution, 1 x Deionized (Rinse) Water (60 ml Each) and Rubber Armour/Stand |
| Accuracy (Temperature) | ±0.05°C |
| Accuracy (mV) | ±1 |
| Accuracy (pH) | ±0.01 |
| Range (pH) | 0 to 14 |
| Resolution (pH) | 0.01 |
| Range (mV) | -1000 to 1000 |
| Resolution (mV) | 1 mV |
| Calibration Points | Up to 5 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction