missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Universal SMPS Power Adapter for Cyberscan 600 meters

The Eutech™ Universal SMPS Power Adapter provides you with the power you need.

Brand:  Thermo Scientific™ 01X030132

Product Code. 11569144

  • 100.20 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Eutech™ Universal SMPS Power Adapter provides you with the power you need. The 100 /240 VAC Power Adapter has US/UK/EUR/Japan plug used for CyberScan 600 series meters.

Features:

  • 9 V, 6 W
  • Used With: CyberScan 600 / 610 / 620 / 650
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Universal SMPS Power Adapter, centre +ve, 100-240V
SMPS Power Adapter
9 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers