Learn More
Thermo Scientific™ Eutech™ USB to IrDA Interface Adapter for Eutech™ 600 Series meters
Click to view available options
For Use With (Equipment):
600 / 610 / 620 / 650 meters
Unit Size:
Each
Description
Eutech™ offers various types of adapters that are available for use with different meters for PC communication. The Eutech™ USB to IrDA Interface Adapter is used to connect Eutech™ 600 Series meters to your computer.
Features:
- Used With: Eutech 600 / 610 / 620 / 650
- Note: This product is not compatible with Windows 10 and 11
Specifications
Specifications
| Description | USB / IrDA Interface Adapter (for 600 Series Meter to PC Communication) |
| Type | USB to IrDA Interface Adapter |
| Compatibility | Not compatible with Windows 10 and 11 |
| For Use With (Equipment) | 600 / 610 / 620 / 650 meters |