Learn More
Heidolph™ Evaporating Cylinder
For use with rotary evaporators
Brand:  Heidolph™ 514-00070-00
 View more versions of this product
Product Code. 10279411
  • 815.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Evaporating cylinders come with a standard NS 29/32 joint size but are also available in NS 24/29
For 500 ml filling capacity.
Specifications
Specifications
Clear
Evaporating Cylinder
Cylinder
Glass
Rotary evaporators
500 mL