Learn More
Heidolph Instruments Evaporating Flask
For use with rotary evaporators
Brand:  Heidolph Instruments 514-73000-00
 View more versions of this product
Product Code. 10457821
  • 87.15 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Evaporating flasks come with a standard NS 29/32 joint size but are also available in NS 24/29
Specifications
Specifications
Borosilicate Glass
Evaporating Flask
500 mL
115/230 VAC