missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Evaporating Flask with Conical Ground Joint

Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks with Conical Ground Joints can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.

Brand:  Lenz Laborglasinstrumente™ 03042937

Product Code. 11826144

  • 182.00 € / Pack of 10
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With center neck and conical joint
  • Pear-shaped with inscription label
  • Available in a range of capacities
Specifications

Specifications

100 mL
Rotary Evaporators
60 mm
Borosilicate Glass, Duran Glass
Center
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lenz Laborglasinstrumente™ Evaporating Flask with Conical Ground Joint > Capacity: 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.