missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Evaporating Flask with Conical Ground Joint
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Evaporating Flasks with Conical Ground Joints can be used to make solutions or to hold, contain, collect or measure chemicals and are manufactured from durable DURAN™ glass.
Brand: Lenz Laborglasinstrumente™ 03042973
This item is not returnable.
View return policy
Description
- With center neck and conical joint
- Pear-shaped with inscription label
- Available in a range of capacities
Specifications
2000 mL | |
Rotary Evaporators | |
158 mm |
Borosilicate Glass, Duran Glass | |
Center |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Evaporating Flask with Conical Ground Joint > Capacity: 2000mL
Spot an opportunity for improvement?Share a Content Correction